Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIBS2 epitope (Integrin beta3 subunit) Mouse anti-Human, Clone: ab62, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT27
Description
Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62 is an antibody against LIBS2 epitope (Integrin beta3 subunit) for use in western blot, flow cytometry.
Specifications
LIBS2 epitope (Integrin beta3 subunit) | |
Monoclonal | |
Unconjugated | |
LIBS2 | |
Human GPIIa-IIIb purified from platelets | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Western Blot | |
ab62 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Mouse | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction