Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light Chain, l Mouse anti-Human, V450, Clone: JDC-12, BD

Catalog No. BDB561379 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561379 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561379 Supplier BD Biosciences Supplier No. 561379
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JDC-12 monoclonal antibody specifically binds to human immunoglobulin light chain, lambda ( λ ). It does not not bind to immunoglobulin κ light chains or heavy chains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Light Chain, l
Applications Flow Cytometry
Classification Monoclonal
Clone JDC-12
Conjugate Violet Fluor 450 (V450)
Description Ig λ; IGL@; IGL; Immunoglobulin lambda locus
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.