Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta-Like Protein 1 Rat anti-Mouse, PE, Clone: 30B11.1, BD

Catalog No. BDB563684 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563684 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563684 Supplier BD Biosciences Supplier No. 563684

Rat Monoclonal Antibody

The 30B11.1 monoclonal antibody specifically binds to Delta-like1 (Dll1) which is also known as Delta1 and DL1. Dll1 is a 90-100 kDa type 1 transmembrane protein. It is a member of the Delta/Serrate/Jagged Family of Notch ligands. Notch ligands are classified by the presence of specific structural motifs including an N-terminal Delta-Serrate-LAG-2 (DSL) domain, EGF repeats, and the DOS domain (a specialized EGF repeat). The Notch family of transmembrane receptors and their ligands control cell-fate “decisions” during the development of many organs in a wide variety of animal species. Dll1 is expressed in the paraxial mesoderm and in some cells in the nervous system in early development. In the adult, Dll1 is expressed in a variety of tissues including at high levels in the heart and at various levels by cells within lymphoid tissues. Although Dll1 does not normally appear to play a role in T cell development, Dll1 and Notch interactions are essential for marginal zone B cell development and reportedly promote the differentiation of antibody producing cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta-Like Protein 1
Applications Flow Cytometry
Classification Monoclonal
Clone 30B11.1
Concentration 0.2mg/mL
Conjugate PE
Description Delta like-1; Delta-like 1; Delta1; Dll1; DL1; DL-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse DL1 Extracellular Domain
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.