Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta-Like Protein 4 Rat anti-Mouse, PE, Clone: 9A1.5, BD

Catalog No. BDB563802 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563802 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563802 Supplier BD Biosciences Supplier No. 563802

Rat Monoclonal Antibody

The 9A1.5 monoclonal antibody specifically binds to an extracellular domain of Delta-Like Protein 4 that is encoded by the Dll4 gene. Delta-Like Protein 4 is a type 1 transmembrane glycoprotein that is also known as Delta-like 4, Delta-like Ligand 4, Delta4 and DL4. It is a member of the Delta/Serrate/Jagged Family of Notch ligands. Notch ligands are classified by the presence of specific structural motifs including an N-terminal Delta-Serrate-LAG-2 (DSL) domain necessary for Notch binding, EGF repeats, and the DOS domain (a specialized EGF repeat). The Notch family of transmembrane receptors and their ligands control cell-fate “decisions” during the development of many organs in a wide variety of animal species. Delta-Like Protein 4 activates cellular signaling pathways by binding to Notch1 and Notch4 receptors. It is involved in embryonic vascular development and tumor angiogenesis, and is induced by vascular endothelial growth factor (VEGF)-A and hypoxia. Delta-Like Protein 4 and Notch receptor signaling are also intimately involved in the regulation of innate and adaptive immunity. Delta-Like Protein 4 is highly expressed by cortical thymic epithelial cells (TEC). Interaction between Delta-Like Protein 4-positive TEC and Notch1 expressed by T cell precursors drives T cell lineage commitment, expansion and maturation within the thymus. Delta-Like Protein 4 is also expressed by dendritic cells and plays a role in peripheral T helper cell differentiation. Blocking of Delta-Like Protein 4 and Notch receptor interactions serves to diminish autoimmune diseases in mouse model systems.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta-Like Protein 4
Applications Flow Cytometry
Classification Monoclonal
Clone 9A1.5
Concentration 0.2mg/mL
Conjugate PE
Description Delta like-4; Delta4; Dll4; DL4; DL-4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse DL4 Extracellular Domain Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.