Learn More
LILRA5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human LILRA5 peptide using ARM Technology.
Supplier: Abnova Corporation H00353514K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family. LIR family members are known to have activating and inibitory functions in leukocytes. Crosslink of this receptor protein on the surface of monocytes has been shown to induce calcium flux and secretion of several proinflammatory cytokines, which suggests the roles of this protein in triggering innate immune responses. This gene is one of the leukocyte receptor genes that form a gene cluster on the chromosomal region 19q13.4. Four alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
LILRA5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
LILRA5 | |
LILRA5 | |
A synthetic peptide of human LILRA5 | |
RUO | |
353514 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5 | |
LILRA5 | |
CD85, CD85F, ILT11, LILRB7, LIR9 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.