Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-LILRB1 Matched Antibody Pair, Abnova™
Supplier: Abnova Corporation H00010859AP51
Includes: Capture antibody: mouse monoclonal anti-LILRB1 (100μg); Detection antibody: rabbit purified polyclonal anti-LILRB1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LILRB1.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
LILRB1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LILRB1. | |
CD85/CD85J/FLJ37515/ILT2/LIR-1/LIR1/MIR-7/MIR7 | |
Mouse, Rabbit | |
RUO | |
10859 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
LILRB1 | |
LILRB1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-LILRB1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-LILRB1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction