Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM-3 Rabbit anti-Human, Mouse, Fish, Frog, Polyclonal, MilliporeSigma™

Catalog No. AB3202MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3202MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3202MI Supplier MilliporeSigma Supplier No. AB3202

Rabbit Polyclonal Antibody

Anti-LIM-3 Antibody is an antibody against LIM-3 for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIM-3
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.
Gene Symbols ABLIM3; KIAA0843; HMFN1661; abLIM-3
Host Species Rabbit
Immunogen C-terminal portion of mouse LIM-3 protein.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014945.2
Target Species Fish, Frog, Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.