Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM Domain Only 2 Mouse anti-Human, Clone: 2E6, MilliporeSigma™

Catalog No. MAB10139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10139MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10139MI Supplier MilliporeSigma Supplier No. MAB10139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect LIM Domain Only 2 using this Anti-LIM Domain Only 2 Antibody, clone 2E6 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIM Domain Only 2
Applications Western Blot
Classification Monoclonal
Clone 2E6
Conjugate Unconjugated
Formulation Liquid containing 0.1% sodium azide.
Gene Accession No. NM_005574.2
Gene Symbols LMO2, RHOM2, TTG2, RBTN2, Rhombotin-2, RBTNL1
Host Species Mouse
Immunogen Recombinant human LMO2
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for one year
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.