Learn More
LIMD1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532475
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is lung tissue.
The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing three LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is downregulated in lung cancer.
Specifications
| LIMD1 | |
| Polyclonal | |
| LIMD1 | |
| LIM domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8994 | |
| 4° C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Q9UGP4 | |
| LIMD1 | |
| Synthetic peptide derived from N-terminus of human LIMD1 protein. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.