Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMK1 Rat anti-Human, Mouse, Clone: 40/00-1B11-1-1, MilliporeSigma™

Catalog No. MAB10750MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10750MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10750MI Supplier MilliporeSigma Supplier No. MAB10750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-LIMK1 Antibody, clone 40/00-1B11-1-1 is validated for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry for the detection of LIMK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMK1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 40/00-1B11-1-1
Conjugate Unconjugated
Formulation Purified in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NM_010717.2
Gene Symbols Limk, Limk1
Host Species Rat
Immunogen GST-tagged recombinant protein corresponding to a region of mouse LIMK1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.