Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lin-28 Mouse anti-Human, PE, Clone: 6D1F9, BD

Catalog No. BDB563398 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563398 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563398 Supplier BD Biosciences Supplier No. 563398

Mouse Monoclonal Antibody

Lin-28 is an evolutionarily conserved RNA-binding protein that is expressed in embryonic tissue, embryonic stem cells, and some adult tissues (e.g. cardiac and skeletal muscle). Within these cells, it is localized to ribosomes, P-bodies, and stress granules. During embryonic development, Lin-28 acts as a “translational enhancer” by binding to certain mRNAs and increasing their translation efficiency. Additionally, Lin-28 is overexpressed in some human cancer cell lines and is associated with the malignant transformation of cells as well as advanced human malignancies. This role is thought to be tied to the binding of Lin-28 to pre-microRNA of let-7, which in its mature form promotes terminal differentiation of stem cells and suppresses tumors. The binding of Lin-28 to the immature version of let-7 blocks its cleavage into mature microRNA, thereby preventing it from carrying out its function in tumor suppression. Lin-28, in combination with additional factors, has been used to reprogram human somatic cells into induced pluripotent stem cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lin-28
Applications Flow Cytometry
Classification Monoclonal
Clone 6D1F9
Conjugate PE
Description Lin-28A, LIN28A, CSDD1, LIN28, ZCCHC1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Lin-28 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.