Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
lin-54 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219357
Description
lin-54 Polyclonal specifically detects lin-54 in Human samples. It is validated for Western Blot.Specifications
| lin-54 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| CXCDC1, JC8.6, MIP120, Protein LIN-54 | |
| Rabbit | |
| 70 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (98%), Bovine (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.82mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6MZP7 | |
| LIN54 | |
| Recombinant protein encompassing a sequence within the center region of human LIN54. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 132660 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction