Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lineage Cocktail Mouse anti-Mouse, Biotin, Clone: 145-2C11, M1/70, RA3-6B2, RB6-8C5, TER-119, BD

Catalog No. BDB559971 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1000 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559971 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559971 Supplier BD Biosciences Supplier No. 559971
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Mouse Lineage Panel has been designed to react with cells from the major hematopoietic cell lineages, such as T lymphocytes, B lymphocytes, monocytes/macrophages, granulocytes, and erythrocytes. This Panel of five pre-diluted biotinylated antibodies is designed for the flow cytometric or immunomagnetic enrichment of hematopoietic progenitors from mouse bone marrow by depletion of cells committed to the T- and B-lymphocytic, myeloid (monocytic and granulocytic), and erythroid lineages. The Panel contains sufficient reagents to stain 10^9 bone marrow cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lineage Cocktail
Applications Flow Cytometry
Classification Monoclonal
Clone 145-2C11, M1/70, RA3-6B2, RB6-8C5, TER-119
Conjugate Biotin
Description N id=test44
Host Species Mouse
Purification Method Affinity Purified
Quantity 1000 Tests
Regulatory Status RUO
Primary or Secondary Primary
Includes Biotin Hamster anti-Mouse CD3e (2mL), Biotin Rat anti-Mouse CD11b (2mL), Biotin Rat anti-Mouse CD45R (2mL), Biotin Rat anti-Mouse Ly-6G and Ly-6C (2mL), Biotin Rat anti-Mouse TER-119/Erythroid cells (2mL)
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.