Learn More
Linear Polyubiquitin Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Linear Polyubiquitin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit Polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | UBC; UB |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to peptide containing the boundary of head-to-tail-linked ubiquitin moieties. see ref, Nat. Cell Biol. 11 123_132, 2009″ |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.