Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07678MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-678-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-678-MI Supplier MilliporeSigma Supplier No. 07678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect LINGO-1 using this Anti-LINGO-1 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINGO-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols LINGO1; LRRN6A; FLJ14594; MGC17422; LERN1; UNQ201
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acid residues 611-620 of human LINGO-1 (APRKFNMKMI)
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_032808.5
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.