Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit LIPIN-1 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3585
Description
Detect LIPIN-1 using this RABBIT ANTI-LIPIN-1 Antibody, N-TERM validated for use in western blotting.
Specifications
Lipin 1 | |
Polyclonal | |
Liquid in Tris-citrate/phosphate buffer, pH 7-8, containing 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Mouse |
Western Blot | |
Unconjugated | |
LPIN1; FLD | |
Synthetic Peptide | |
100 μg | |
NM_015763.3 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction