Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liver receptor homolog-1 Mouse anti-Mouse, Clone: 1LRH-2E1, MilliporeSigma™

Catalog No. MABE215MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE215MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE215MI Supplier MilliporeSigma Supplier No. MABE215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-LRH-1 Antibody, clone 1LRH-2E1 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect LRH-1 also known as Liver receptor homolog 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Liver receptor homolog-1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1LRH-2E1
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2aλ ascites with 0.05% sodium azide.
Gene Accession No. NP_109601
Gene Symbols Nr5a2, Lrh1
Host Species Mouse
Immunogen Linear peptide corresponding to the mouse Liver receptor homolog-1.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.