Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKB1 Mouse anti-Human, Mouse, Rat, Clone: 5c10, MilliporeSigma™

Catalog No. 05832MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-832-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-832-MI Supplier MilliporeSigma Supplier No. 05832
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-LKB1 Antibody, clone 5c10 is validated for use in immunocytochemistry, western blotting for the detection of LKB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LKB1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5c10
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% Sodium Azide, pH 7.4.
Gene Accession No. NM_000455
Gene Symbols STK11, PJS, LKB1
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 1-433 of human LKB1
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.