Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lethal giant larvae homolog 1 (Drosophila), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004194 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-194 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-194 Supplier Abnova Corporation Supplier No. H00003996A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LLGL1.

This gene encodes a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq

Sequence: GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSP

Specifications

Antigen lethal giant larvae homolog 1 (Drosophila)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LLGL1.
Formulation 50% glycerol
Gene LLGL1
Gene Accession No. NM_004140
Gene Alias DLG4/HUGL/HUGL-1/HUGL1/LLGL
Gene Symbols LLGL1
Host Species Mouse
Immunogen LLGL1 (NP_004131, 911 a.a. ∼ 1010 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3996
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.