Learn More
LIM and cysteine-rich domains 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene contains a cysteine-rich domain in the N-terminal region and 2 LIM domains in the C-terminal region. It also has several potential phosphorylation and N-myristoylation sites and a single potential N-glycosylation site. The presence of LIM domains implies involvement in protein-protein interactions. Expression of this gene has been detected in most tissues, with highest expression in skeletal muscle. Transcript variants utilizing alternative polyA signals have been observed. [provided by RefSeq
Specifications
Specifications
| Antigen | LIM and cysteine-rich domains 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant LMCD1. |
| Formulation | 50% glycerol |
| Gene | LMCD1 |
| Gene Accession No. | NM_014583 |
| Gene Symbols | LMCD1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.