Learn More
Description
LMO1 enocdes a cysteine-rich, two LIM domain transcriptional regulator. It is mapped to an area of consistent chromosomal translocation in chromosome 11, disrupting it in T-cell leukemia, although more rarely than the related gene, LMO2 is disrupted. [provided by RefSeq
Specifications
Specifications
| Antigen | LIM domain only 1 (rhombotin 1) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1A9 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant LMO1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LMO1 |
| Gene Accession No. | NM_002315 |
| Gene Alias | MGC116692/RBTN1/RHOM1/TTG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.