Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMX-1 Rabbit anti-Mouse, Rat, Hamster, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB10533
Description
Anti-LMX-1 Antibody is an antibody against LMX-1 for use in western blotting.
Specifications
| LMX-1 | |
| Polyclonal | |
| Unpurified rabbit polyclonal serum containing 0.05% sodium azide. | |
| Rabbit | |
| Unpurified | |
| Primary | |
| Hamster, Mouse, Rat | |
| Serum |
| Western Blot | |
| Unconjugated | |
| LMX-12; LMX1.1; LMX-1.1; LMX1 | |
| GST-tagged recombinant protein corresponding to the C-terminus of hamster LMX-1. | |
| 100 μL | |
| AAB62320 | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction