Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-780 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-780 Supplier Abnova Corporation Supplier No. H00129607A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CMPK2.

Mitochondrial UMP-CMP kinase (EC 2.7.2.14) is a component of the salvage pathway for nucleotide synthesis. Other enzymes of the salvage pathway include thymidine kinase-2 (TK2; MIM 188250), deoxynucleotidase-2 (NT5M; MIM 605292), deoxyguanosine kinase (DGUOK; MIM 601465), adenylate kinase-2 (AK2; MIM 103020), adenylate kinase-3 (AK3; MIM 609290), adenylate kinase-3-like-1 (AK3L1; MIM 103030), and nucleoside diphosphate kinase (NME4; MIM 601818) (Xu et al., 2008 [PubMed 17999954]).[supplied by OMIM

Sequence: GACQEAPRPHLGEFEADPRGQLWQRLWEVQDGRRLQVGCAQVVPVPEPPLHPVVPDLPSSVVFPDREAARAVLEECTSFIPEARAVLDLVDQCPKQIQKG

Specifications

Antigen cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CMPK2.
Formulation 50% glycerol
Gene CMPK2
Gene Accession No. NM_207315
Gene Alias TYKi/UMP-CMPK2
Gene Symbols CMPK2
Host Species Mouse
Immunogen CMPK2 (NP_997198, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 129607
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.