Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LON peptidase N-terminal domain and ring finger 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human LONRF2 protein.
Supplier: Abnova Corporation H00164832B01
Description
Sequence: MKNYEQALQDASAACQNEPLLIKGHQVKAQALSGLGRSKEVLKEFLYCLALNPECNSVKKEAQKVMCEVLFSATANVHENLTSSIQSRLKAQGHSHMNAQALLEEGDAGSSENSSEKSDMLGNTNSSVLYFILGLHFEEDKKALESILPTAPSAGLKRQFPDDVEDAPDLNAPGKIPKKDLSPQRSPNSETEESQGLSLDVTDFECALCMRLLFEPVTTPCGHTFCLKCLERCLDHAPHCPLCKDKLSELLASRNFNITVLAEELIFRYLPDELSDRKRIYDEEMSELSNLTRDVPIFVCAMAFPTVPCPLHVFEPRYRLMIRRCMETGTKRFGMCLSAEHAGLSEYGCMLEIKDVRTFPDGSSVVDAIGISRFRVLSHRHRDGYNTADIEYLEDEKVEGPEYEELAALHDSVHQQSVSWFASLQDRMKEQILSHFGVMPDREPEPQSNPSGPAWSWWILAVLPLERKAQLAILGMTSLKERLLAIRRILVIITRKMNSRQELANARERNNSpecifications
LON peptidase N-terminal domain and ring finger 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human LONRF2 protein. | |
LONRF2 | |
FLJ45273/MGC126711/MGC126713/RNF192 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC101664.1 | |
LONRF2 | |
LONRF2 (AAI01665.1, 1 a.a. ∼ 511 a.a) full-length human protein. | |
RUO | |
Yes | |
164832 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction