Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Low Density Lipoprotein Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3230MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3230MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3230MI Supplier MilliporeSigma Supplier No. AB3230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Low Density Lipoprotein Antibody, copper oxidized is an antibody against Low Density Lipoprotein for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Low Density Lipoprotein
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 100μL of sterile distilled water.
Host Species Rabbit
Immunogen Fully Cu2+-oxidized LDL.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.