Learn More
lysyl oxidase-like 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LOXL3.
Supplier: Abnova Corporation H00084695A01
Description
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family. Alternatively spliced transcript variants of this gene have been reported but their full-length nature has not been determined. [provided by RefSeq
Sequence: IRPAVGWGRRPLPVTEGLVEVRLPDGWSQVCDKGWSAHNSHVVCGMLGFPSEKRVNAAFYRLLAQRQQHSFGLHGVACVGTEAHLSLCSLEFYRANDTARSpecifications
lysyl oxidase-like 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant LOXL3. | |
LOXL3 | |
LOXL | |
Mouse | |
50 μL | |
Endocrine/Metabolism | |
Primary | |
Human, Mouse | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_032603 | |
LOXL3 | |
LOXL3 (NP_115992, 171 a.a. ∼ 270 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
84695 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.