Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPA receptor 3 (Edg-7) Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT113
Description
Detect LPA receptor 3 (Edg-7) using this Anti-LPA receptor 3 (Edg-7) Antibody validated for use in Western Blotting, immunohistochemistry (paraffin).
Specifications
LPA receptor 3 (Edg-7) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Lpar3; Edg7; Lpa3 | |
KLH-conjugated linear peptide corresponding to the extracellular domain of human LPA receptor 3. | |
100 μg | |
Np_036284 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction