Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAM-1 Rat anti-Mouse, PE, Clone: DATK32, BD

Catalog No. BDB553811 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553811 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553811 Supplier BD Biosciences Supplier No. 553811
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The DATK32 antibody reacts with an epitope specific to the α4β7 integrin heterodimer. α4β7 (LPAM-1) is expressed on most mature lymphocytes and on small subsets of thymic and bone marrow cells. It interacts with several ligands, including VCAM-1 (CD106), fibronectin, and MAdCAM-1. DATK32 antibody induces α4β7-dependent lymphocyte aggregation, but it inhibits other α4β7- mediated lymphocyte adhesion events, including binding to fibronectin, MAdCAM-1, and VCAM-1 (CD106).

Host Species: Rat
Clone: DATK32
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: AKR/Cum mouse T lymphoma line TK1

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAM-1
Applications Flow Cytometry
Classification Monoclonal
Clone DATK32
Concentration 0.2mg/mL
Conjugate PE
Description Itga4Itgb7; α4β7 Integrin; alpha 4 beta 7 integrin; CD49dβ7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen AKR/Cum mouse T lymphoma line TK1
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.