Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LR11 Mouse, Unlabeled, Clone: 48, BD

Catalog No. BDB611860 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611860 50 μg
BDB611861 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611860 Supplier BD Biosciences Supplier No. 611860
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The low-density lipoprotein receptor (LDLR) functions in lipoprotein transport pathways, and is involved in familial hypercholesterolemia. Homologues of the LDLR may have diverse functions and their ligands may include biological inactive plasma carrier complexes, plasma lipoproteins, yolk precursors, toxins, and extracellular lipoproteins. LR11 (also known as sorLA-1 and gp250) is a lipoprotein receptor homologue that contains 11 LDL receptor ligand binding repeats (LDLRs), 5 LDL receptor “YWTD”repeats, a large fibronectin-type III (FIII) hexarepeat domain similar to neural adhesion proteins, and a domain with similarity to the yeast receptor for vacuolar protein sorting (Vsp10p). LR11mRNA is expressed at high levels in brain, but is also found in liver, pancreas, adrenal gland, and testis. LR11 can bind the ER and Golgi localized receptor associated protein (RAP), which binds to many members of the LDLR family and prevents aggregation of ligands to the LDLRs. In addition, LR11 binds apolipoprotein E-containing lipoproteins. Thus, LR11 is a lipoprotein binding receptor that may have additional functions related to protein sorting and neuronal development.

Host Species: Mouse
Clone: 48
Isotype: IgG2a
Species Reactivity [for Features Main]: Rat
Immunogen: Human LR11 aa. 1220-1337

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen LR11
Applications Western Blot
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate Unconjugated
Description SorLA; gp250; Lipoprotein Receptor-11
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human LR11 aa. 1220-1337
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.