Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LRP-2/Megalin Clone: 10D5.1,

Catalog No. MABS489MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS489MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS489MI Supplier MilliporeSigma™ Supplier No. MABS489
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects LRP-2/Megalin Clone: 10D5.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Low-density lipoprotein receptor-related protein 2 (UniProt P98164; also known as Calcium sensor protein, Glycoprotein 330, gp330, Heymann nephritis antigen homolog, LRP-2, Megalin) is encoded by the LRP2 (also known as DBS) gene (Gene ID 4036) in human. Megalin/LRP-2 is a 600kDa type I membrane receptor initially produced with a signal peptide sequence (a.a. 1-25). The mature receptor has a large extracellular region (a.a. 26-4423), a transmembrane segment (a.a. 4424-4446), and cytoplasmic tail (a.a. 4447-4655). Megalin belongs to the LDLR family of membrane glycoproteins whose members also inculde the low-density lipoprotein receptor (LDLR), the very-low-density lipoprotein (VLDL) receptor, the LDLR-related protein (LRP), and the apolipoprotein E receptor 2 (ApoER2). The overall extracellular structural features are similar among all LDLR family members. Megalin contains four clusters of cysteine-rich Complement-type/LDLR Class A repeats that mediate ligand-binding. These are separated by 17 EGF-like repeats and eight cysteine-poor spacer regions that contain YWTD motifs involved in the pH-dependent ligands dissociation in endosomal compartments. Megalin is a multiligand, endocytic receptor expressed primarily in the apical membranes of polarized epithelial cells. In addtion to functioning alone, megalin is also reported to interact with another structurally distinct endocytic receptor cubilin in several tissues where they are co-expressed. The internalization of several cubilin ligands is reported to be strongly facilitated in the presence of megalin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-2/Megalin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10D5.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P98164
Gene Symbols LRP2, DBS
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the LDL-receptor class A repeats 19-21 of human LRP-2/Megalin.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004516
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.