Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LRP, Light Chain Clone: 5A6,

Catalog No. 438192100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
43-819-2100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-819-2100UG Supplier MilliporeSigma™ Supplier No. 438192100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects LRP, Light Chain Clone: 5A6 in Human, Mouse, Rabbit samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

LRP (Low Density Lipoprotein Receptor-Related Protein) contributes to the pathology of Alzheimer's disease by influencing both the production and clearance of the β-amyloid peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP, Light Chain
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A6
Conjugate Unconjugated
Formulation In 100mM NaCl, 50mM sodium phosphate, 1mM EDTA, pH 6.6.
Host Species Mouse
Immunogen purified, human placenta LRP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽85kDa LRP protein.
Target Species Human, Mouse, Rabbit
Content And Storage −20°C
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.