Learn More
low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, Mouse, Clone: 3H2, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes an apolipoprotein E receptor, a member of the low density lipoprotein receptor (LDLR) family. Apolipoprotein E is a small lipophilic plasma protein and a component of lipoproteins such as chylomicron remnants, very low density lipoprotein (VLDL), and high density lipoprotein (HDL). The apolipoprotein E receptor is involved in cellular recognition and internalization of these lipoproteins. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | low density lipoprotein receptor-related protein 8, apolipoprotein e receptor |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3H2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant LRP8. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LRP8 |
| Gene Accession No. | NM_004631 |
| Gene Alias | APOER2/HSZ75190/MCI1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.