Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC25 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524062
Description
May be involved in the activation of cells of innate and acquired immunity.
Specifications
LRRC25 | |
Polyclonal | |
LRRC25 | |
MAPA, leucine-rich repeat-containing protein 25, monocyte and plasmacytoid activated molecule, monocyte and plasmacytoid-activated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126364 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q8N386 | |
LRRC25 | |
KLH conjugated synthetic peptide between 242-270 amino acids from the C-terminal region of human LRRC25 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction