Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002927 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-927 Supplier Abnova Corporation Supplier No. H00079705D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human LRRK1 protein.

Sequence: MSQRPPSMYWCVGPEESAVCPERAMETLNGAGDTGGKPSTRGGDPAARSRRTEGIRAAYRRGDRGGARDLLEEACDQCASQLEKGQLLSIPAAYGDLEMVRYLLSKRLVELPTEPTDDNPAVVAAYFGHTAVVQELLESLPGPCSPQRLLNWMLALACQRGHLGVVKLLVLTHGADPESYAVRKNEFPVIVRLPLYAAIKSGNEDIAIFLLRHGAYFCSYILLDSPDPSKHLLRKYFIEASPLPSSYPGKTVSSHCLWSVF

Specifications

Antigen LRRK1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human LRRK1 protein.
Formulation No additive
Gene LRRK1
Gene Accession No. BC068080.1
Gene Alias FLJ23119/FLJ27465/KIAA1790/RIPK6/Roco1
Gene Symbols LRRK1
Host Species Rabbit
Immunogen LRRK1 (AAH68080.1, 1 a.a. ∼ 261 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79705
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.