Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK2 Mouse anti-Human, Mouse, Clone: N138/6, MilliporeSigma™

Catalog No. MABN40MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN40MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN40MI Supplier MilliporeSigma Supplier No. MABN40
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LRRK2 Antibody, clone N138/6 detects level of LRRK2 and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRK2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N138/6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_940980
Gene Symbols LRRK2, PARK8, AURA17, RIPK7, ROCO2
Host Species Mouse
Immunogen Recombinant protein corresponding to human LRRK2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.