Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSD1 Mouse anti-Human, Clone: 3D12, MilliporeSigma™

Catalog No. MABE114MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE114MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE114MI Supplier MilliporeSigma Supplier No. MABE114

Mouse Monoclonal Antibody

Use Anti-LSD1 Antibody, clone 3D12 (Mouse Monoclonal Antibody) validated in western blotting to detect LSD1 also known as BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSD1
Applications Western Blot
Classification Monoclonal
Clone 3D12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001009999
Gene Symbols BHC110, KDM1, LSD1
Host Species Mouse
Immunogen Full length recombinant protein corresponding to human LSD1.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.