Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABE365MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE365MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE365MI Supplier MilliporeSigma Supplier No. ABE365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-LSD1 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation to detect LSD1 also known as Lysine-specific histone demethylase 1A, BRAF35-HDAC complex protein BHC110.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSD1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols KDM1A; AOF2; KDM1; LSD1
Host Species Rabbit
Immunogen Histidine-tagged recombinant protein corresponding to full length LSD1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_055828
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.