Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSF Mouse, Unlabeled, Clone: 14, BD

Catalog No. BDB610818 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610818 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610818 Supplier BD Biosciences Supplier No. 610818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSF (αCP2 or LBP-1c) is a transcription factor originally identified by its binding to, and induction of, the SV40 late promoter. LSF displays some similarity to the Elf-1/NTF transcription factors of Drosophila. It is primarily found in

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen LSF
Applications Western Blot
Classification Monoclonal
Clone 14
Concentration 250μg/mL
Conjugate Unconjugated
Description αCP2; LBP-1c
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human LSF aa. 205-414
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.