Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSm1 Rat anti-Mouse, Clone: 5F3, MilliporeSigma™

Catalog No. MABE893MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE893MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE893MI Supplier MilliporeSigma Supplier No. MABE893
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-LSm1 Antibody, clone 5F3 is an antibody against LSm1 for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSm1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5F3
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_080308
Gene Symbols Lsm1
Host Species Rat
Immunogen Recombinant protein corresponding to mouse LSm1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.