Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LST1 Rat anti-Human, Clone: 7E2, MilliporeSigma™

Catalog No. MABF242MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF242MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF242MI Supplier MilliporeSigma Supplier No. MABF242
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-LST1 antibody, clone 7E2 detects level of LST1 and has been published and validated for use in LST1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LST1
Applications Western Blot
Classification Monoclonal
Clone 7E2
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001160010
Gene Symbols LST1, B144
Host Species Rat
Immunogen His-tagged recombinant protein corresponding to human LST1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.