Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LT-α Mouse anti-Human, Biotin, Clone: 359-81-11, BD

Catalog No. BDB554555 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554555 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554555 Supplier BD Biosciences Supplier No. 554555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 359-81-11 antibody reacts with human Lymphotoxin-α (LT-α, also known as tumor necrosis factor-β or TNF-β). The immunogen used to generate the 359-81-11 hybridoma was recombinant human LT-α. This is a neutralizing antibody.

Host Species: Mouse
Clone: 359-81-11
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human LT-α (TNF-β)

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen LT-α
Applications ELISA
Classification Monoclonal
Clone 359-81-11
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human LT-α (TNF-β)
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.