Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-LTA, Flag/His, Clone: 3B5, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation H00004049M09
Specifications
| LTA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| LTA | |
| LT/TNFB/TNFSF1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 3B5 | |
| Mouse monoclonal antibody raised against a partial recombinant LTA. | |
| PBS with no preservative; pH 7.2 | |
| BC034729.1 | |
| LTA | |
| Recombinant Flag/His fusion protein corresponding to amino acids 58-205 of human LTA. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction