Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LTA, Flag/His, Clone: 4C7, Abnova™

Catalog No. 89300170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-170 Supplier Abnova Corporation Supplier No. H00004049M03
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen LTA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LTA.
Dilution The optimal working dilution should be determined by the end user.
Formulation PBS with no preservative; pH 7.2
Gene LTA
Gene Accession No. BC034729.1
Gene Alias LT/TNFB/TNFSF1
Gene Symbols LTA
Host Species Mouse
Immunogen Recombinant Flag/His fusion protein corresponding to amino acids 58-205 of human LTA.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4049
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.