Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTBP4, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-527 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-527 Supplier Abnova Corporation Supplier No. H00008425K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human LTBP4 peptide using ARM Technology.

Transforming growth factor-beta, or TGFB (see TGFB1; MIM 190180), remains biologically latent after secretion, blocked from activity by its N-terminal propeptide (the latency-associated peptide, or LAP). TGFB is activated by its dissociation from LAP. The latent form of TGFB is secreted in association with a latent TGFB-binding protein, or LTBP (see LTBP1; MIM 150390), and assembled into the extracellular matrix. LTBPs are mainly composed of epidermal growth factor (EGF; MIM 131530)-like repeats and protein domains with 8-cysteine repeats, both of which are also found in fibrillins (e.g., MIM 134797). See Oklu and Hesketh (2000) [PubMed 11104663] for a review of the LTBP gene family.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen LTBP4
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description latent transforming growth factor beta binding protein 4
Formulation In 1X PBS, pH 7.4
Gene LTBP4
Gene Accession No. LTBP4
Gene Alias FLJ46318, FLJ90018, LTBP-4, LTBP-4L
Gene Symbols LTBP4
Host Species Rabbit
Immunogen A synthetic peptide of human LTBP4
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8425
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.