Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTBR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA130176
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA130176 50 μg
1 options

Catalog No. PIPA130176

Supplier: Thermo Scientific PA130176

LTBR Antibody, PA1-30176, from Invitrogen™. Species Reactivity: Human; Applications: ELISA, Flow Cytometry, Western Blot.

LTBR Antibody, PA1-30176, from Invitrogen. Species Reactivity: Human; Applications: ELISA, Flow Cytometry, Western Blot.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
Host Species: Goat
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant human lymphotoxin betaR extracellular domain, expressed in mouse NSO cells.
Formulation: 50μg purified antibody (0.1mg/mL) in PBS with 5% trehalose and no preservative
Synonyms: CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFRSF3
Uniprot Accession: P36941
Entrez GeneID: 4055
Gene Symbol: LTBR
PA1-30176 detects LTBR from human samples. PA1-30176 has been successfully used in flow cytometry, Western blot applications. The PA1-30176 immunogen is: Recombinant human lymphotoxin betaR extracellular domain, expressed in mouse NSO cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Accession No. P36941
Gene Alias CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFRSF3
Gene Symbols LTBR
Host Species Goat
Immunogen Recombinant human lymphotoxin betaR extracellular domain, expressed in mouse NSO cells.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.