Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase Goat anti-Invertebrates, Polyclonal, MilliporeSigma™

Catalog No. AB3256100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3256100UL 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3256100UL Supplier MilliporeSigma Supplier No. AB3256100UL
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Anti-Luciferase Antibody, Firefly detects level of Luciferase and has been published and validated for use in ELISA, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luciferase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoglobulin purified by delipidation, salt fractionation and ion exchange chromatography. Liquid in 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% sodium azide.
Host Species Goat
Immunogen Luciferase [Photinus pyralis (Firefly)].
Quantity 100 μL
Primary or Secondary Primary
Target Species Invertebrate
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.