Learn More
MilliporeSigma™ anti-Luteinizing Hormone/Choriogonadotropin Receptor Ab 2, Polyclonal,
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Antigen | Luteinizing Hormone/Choriogonadotropin Receptor |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide. |
| Gene Symbols | LHB; hLHB; LSH-B; LH-B; LSH-beta; CGB4 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the N-terminal extracellular domain of human Luteinizing Hormone/Choriogonadotropin [LH/CG] Receptor. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.