Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luteinizing Hormone Releasing Hormone Rabbit anti-Chicken, Human, Mouse, Rat, Rabbit, Sheep, Fish, Polyclonal, MilliporeSigma™

Catalog No. AB1567MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1567MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1567MI Supplier MilliporeSigma Supplier No. AB1567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Luteinizing Hormone Releasing Hormone Antibody detects level of Luteinizing Hormone Releasing Hormone and has been published and validated for use in immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luteinizing Hormone Releasing Hormone
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit antisera. Liquid
Gene Symbols GNRH1; LHRH; LNRH; GRH; GNRH
Host Species Rabbit
Immunogen LHRH (Sigma) conjugated to thyroglobulin.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000825.3; NM_001083111.1
Target Species Chicken, Fish, Human, Mouse, Rabbit, Rat, Sheep
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.