Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49D Rat anti-Mouse, Unlabeled, Clone: 4E5, BD

Catalog No. BDB555312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555312 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555312 Supplier BD Biosciences Supplier No. 555312
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4E5 antibody reacts with Ly-49D, which is expressed on subsets of natural killer (NK) cells in C57BL/6, C3H/He (at a very low frequency), and SJL, but not DBA/2, AKR, CBA/J, or BALB/c mice. Unlike Ly-49D antigen has not been betected on NK-1.1+ (or DX5+) T cells. In 129/J mice, the 4E5 antibody cross-reacts with Ly-49O, Ly-49R, and Ly-49V. The Ly-49 family of NK-cell receptors, members of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains, which bind to MHC class I alloantigens. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. Ly-49D weakly binds to MHC class I antigens of the k halpotype, and Ly-49D+ IL-2-activated NK cells lyse target cells expressing H-2[a], H-2[b], H-2[d], H-2[k], H-2[p], H-2[q], and H-2[s] and the CHO (Chinese hamster ovary) cell line. Ly-49D+ cells mediate allogenic resistance to H-2d bone marrow transplantation. In vitro studies suggest that the Ly-49D receptor mediates activation of NK-cell cytolytic activity via tyrosine phosphorylation of their ITIMs (Immunoreceptor Tyrosine-based Inhibitory Motifs). Molecular differences between the Ly-49D stimulatory receptor and the inhibitory members of the Ly-49 family include the absence of an ITIM in Ly-49D, the lack of phosphorylation of Ly-49D in activated NK cells, and the association of a novel tyrosine-phosphorylated protein (pp16) with Ly-49D in activated NK cells. Ly-49O and Ly-49V are closely related to Ly-49A[B6] and, like Ly-49A, have ITIM domains. Ly-49O- and Ly-49V-transfected 293T (human kidney epithelial) cells bind tetramers of H-2D[b], D[b], D[k], and L[d]. In addition, the Ly-49V-transfected cells also bind K[b], K[d], and K[k]. Ly-49R is closely related to Ly-49D[B6] and is putative activating receptor due to its lack of an ITIM domain. Ly-49R-transfected 293T cells bind soluble tetramers of H-2D[b], D[d], D[k], and L[d].

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49D
Applications Flow Cytometry
Classification Monoclonal
Clone 4.00E+05
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Not reported
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.