Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-51 Rat anti-Mouse, Biotin, Clone: 6C3, BD

Catalog No. BDB553159 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553159 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553159 Supplier BD Biosciences Supplier No. 553159
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6C3 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP

Host Species: Rat
Clone: 6C3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: C57L mouse Pre-B lymphhoma cell line L1-2

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-51
Applications Flow Cytometry
Classification Monoclonal
Clone 6C3
Concentration 0.5mg/mL
Conjugate Biotin
Description 6C3/BP-1 Antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen C57L mouse Pre-B lymphhoma cell line L1-2
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.